Learn More
Chemglass Life Sciences Spatula, Stainless Steel #304, 12" OAL, Non-Magnetic, Tapered End
SDP
Catalog No. CG198212
Encompass_Preferred
Spatula will pass through a 14/10 or 14/20 outer joint. Made of non-magnetic stainless steel having a mirror finish. One end is rounded, the other is tapered to 1/8in (3.3mm). Both ends are approx. 5/16in (8mm) wide x 2in (51mm) in length.
Catalog No. CG198212 Supplier Chemglass Life Sciences Supplier No. CG198212
Spatula will pass through a 14/10 or 14/20 outer joint. Made of non-magnetic stainless steel having a mirror finish. One end is rounded, the other is tapered to 1/8in (3.3mm). Both ends are approx. 5/16in (8mm) wide x 2in (51mm) in length.