Learn More
Foxx Life Sciences EZBio Stainless Steel Spatula Polished with Steel Handle, 304 SS, 100mm, 1/EA
Catalog No. NC1795124
Encompass_Preferred
Foxx Life Sciences Stainless Steel Spatulas are ergonomically designed for ease of use and the versatility of the product makes it possible to support numerous processes, such as formulation work, measuring, sample gathering, powder prep, granulate work, with a variety of media including pastes and liquids. Manufactured with a single piece of high-quality 304 Stainless Steel, they are free of edges, crevices, or undercuts, to eliminate the possibility of cross contamination, product residue, or cleaning agent deposits. They are perfect for uses in chemistry, pharmaceuticals, cosmetics, and food. These spatulas are available in 100mm and 150mm sizes.304 Stainless Steel High-gloss polishedEasy to clean hygienic design.
Catalog No. NC1795124 Supplier Foxx Life Sciences Supplier No. 25C3611FLS
Foxx Life Sciences Stainless Steel Spatulas are ergonomically designed for ease of use and the versatility of the product makes it possible to support numerous processes, such as formulation work, measuring, sample gathering, powder prep, granulate work, with a variety of media including pastes and liquids. Manufactured with a single piece of high-quality 304 Stainless Steel, they are free of edges, crevices, or undercuts, to eliminate the possibility of cross contamination, product residue, or cleaning agent deposits. They are perfect for uses in chemistry, pharmaceuticals, cosmetics, and food. These spatulas are available in 100mm and 150mm sizes.304 Stainless Steel High-gloss polishedEasy to clean hygienic design.