Learn More
Hach Company Spatula, Sterile Sampler
Supplier:  Hach Company 2600800
Encompass
Avoid sample contamination. Use this disposable, sterile spatula, 229 mm (9") long, with permanent handle. Holds abundant volume (36 cc when level full) to save time. Ideal for taking samples from bags. Individually wrapped. Bel-Art.
Catalog No. 50-199-6308