Learn More
World Precision Instrument Wecker Spatula, 14cm, 3mm wide blade
SDP
Catalog No. 50822814
Encompass
A Wecker Spatula is a surgical instrument designed for ophthalmology to lift, separate, dissect or manipulate delicate tissues of the eye like the conjunctiva or cornea. Named after the German ophthalmologist Professor Ernst Wecker, the Wecker Spatula has a fine, narrow blade. Our Wecker Spatula is 14 cm (5.5") long and has a solid square handle for a secure grip. It comes with either a 2 mm or 3 mm wide, flat blade with a rounded tip. One is made of surgical grade stainless steel, and the other is manufactured from premium German steel. Both can be sterilized with a chemical sterilant or by autoclaving. Always clean your surgical instruments promptly after use to ensure they last a long time.
Catalog No. 50-822-814 Supplier World Precision Instrument Supplier No. 501772
A Wecker Spatula is a surgical instrument designed for ophthalmology to lift, separate, dissect or manipulate delicate tissues of the eye like the conjunctiva or cornea. Named after the German ophthalmologist Professor Ernst Wecker, the Wecker Spatula has a fine, narrow blade. Our Wecker Spatula is 14 cm (5.5") long and has a solid square handle for a secure grip. It comes with either a 2 mm or 3 mm wide, flat blade with a rounded tip. One is made of surgical grade stainless steel, and the other is manufactured from premium German steel. Both can be sterilized with a chemical sterilant or by autoclaving. Always clean your surgical instruments promptly after use to ensure they last a long time.