Learn More
Fine Science Tools SPATULAE
SDP
Catalog No. NC9222532
Encompass
Individual Item Name: Iris Spatulae - Straight/13cm/ Tip Shape: Straight/ Tip Dimensions: 33 x 3 x 0.4mm/ Alloy: Stainless Steel/ Length: 13cm
Catalog No. NC9222532 Supplier Fine Science Tools Supplier No. 1009413
Individual Item Name: Iris Spatulae - Straight/13cm/ Tip Shape: Straight/ Tip Dimensions: 33 x 3 x 0.4mm/ Alloy: Stainless Steel/ Length: 13cm