Learn More
Selleck Chemical LLC Spautin-1 10mg 1262888-28-7
SDP
Catalog No. 501365375
Encompass_Preferred
Spautin-1 is a potent and specific autophagy inhibitor, and inhibits the deubiquitinating activity of USP10 and USP13 with IC50 of ?0.6-0.7 ?M. Spautin-1 enhances apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5375 Supplier Selleck Chemical LLC Supplier No. S788810MG
Spautin-1 is a potent and specific autophagy inhibitor, and inhibits the deubiquitinating activity of USP10 and USP13 with IC50 of ?0.6-0.7 ?M. Spautin-1 enhances apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.