Learn More
Medchemexpress LLC HY-12990 10mg , Spautin-1 CAS:1262888-28-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1299010MG
Encompass_Preferred
HY-12990 10mg Spautin-1 CAS: 1262888-28-7 Spautin-1 is a specific and potent autophagy inhibitor which inhibits ubiquitin-specific peptidases, USP10 and USP13 with IC50s of 0.6-0.7 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9673