Learn More
Medchemexpress LLC Spautin-1 | 1262888-28-7 | 99.5% | 100 MG
SDP
Catalog No. 5000339902
Encompass_Preferred
Spautin-1 is a specific and potent autophagy inhibitor that targets ubiquitin-specific peptidases, USP10 and USP13. It is used in research for its role in enhancing apoptosis and ameliorating acute pancreatitis. This product comes as a white to light yellow solid.
  • Inhibits ubiquitin-specific peptidases, USP10 and USP13
  • Enhances apoptosis in CmL cells
  • Pro-apoptotic activity associated with activation of GSK3β
  • Increases cytotoxicity in K562 cells
  • Ameliorates acute pancreatitis
  • Diminishes elevated serum amylase and lipase levels
  • Inhibits increased serum TNFα levels
  • Reduces inflammation damage like edema, degeneration, and necrosis
Catalog No. 50-003-39902 Supplier Medchemexpress LLC Supplier No. HY12990100MG
Spautin-1 is a specific and potent autophagy inhibitor that targets ubiquitin-specific peptidases, USP10 and USP13. It is used in research for its role in enhancing apoptosis and ameliorating acute pancreatitis. This product comes as a white to light yellow solid.
  • Inhibits ubiquitin-specific peptidases, USP10 and USP13
  • Enhances apoptosis in CmL cells
  • Pro-apoptotic activity associated with activation of GSK3β
  • Increases cytotoxicity in K562 cells
  • Ameliorates acute pancreatitis
  • Diminishes elevated serum amylase and lipase levels
  • Inhibits increased serum TNFα levels
  • Reduces inflammation damage like edema, degeneration, and necrosis