Learn More
Perkin Elmer US LLC SPB 15-40, 115 V/230 V
Supplier:  Perkin Elmer US LLC N9300800
Encompass_Preferred
The SPB series of block digestion systems include graphite blocks that are PTFE-coated to resist aggressive corrosive attack for guaranteed long life in harsh laboratory environments.
Catalog No. 50-210-4237