Learn More
Perkin Elmer US LLC SPB Probe 6 in. for 50 mL Tubes
Catalog No. 502103721
Encompass_Preferred
The SPB Probe provides direct control and monitoring of actual sample temperature via a corrosion-resistant, PFA-coated temperature probe. Each probe is supplied with five Probe Watch Glasses and a probe holder.
Catalog No. 50-210-3721 Supplier Perkin Elmer US LLC Supplier No. N9308018
The SPB Probe provides direct control and monitoring of actual sample temperature via a corrosion-resistant, PFA-coated temperature probe. Each probe is supplied with five Probe Watch Glasses and a probe holder.