Learn More
Enzo Life Sciences Sphingosylphosphorylcholine (10 mg). CAS: 82970-80-7
SDP
Catalog No. 502012117
Encompass_Preferred
Sphingosylphosphorylcholine induces release of Ca2+ from intracellular stores via an IP3 independent pathway in permeabilized pancreatic acinar cells. It is an extremely potent mitogen in numerous cell types and induces neurite outgrowth in mouse neuroblastoma cells. It rapidly activates transcription factor AP-14 and transiently activates MAP kinase. Purity: ≥98% (HPLC). Solubility: Soluble in water (>50 mg/ml) or ethanol (>50 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2117 Supplier Enzo Life Sciences Supplier No. BMLSL1410010
Sphingosylphosphorylcholine induces release of Ca2+ from intracellular stores via an IP3 independent pathway in permeabilized pancreatic acinar cells. It is an extremely potent mitogen in numerous cell types and induces neurite outgrowth in mouse neuroblastoma cells. It rapidly activates transcription factor AP-14 and transiently activates MAP kinase. Purity: ≥98% (HPLC). Solubility: Soluble in water (>50 mg/ml) or ethanol (>50 mg/ml). Long Term Storage: -20°C.