Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ SPC™ Absorbents HAZWIK™ Quick Response Kits

Catalog No. p-9510008 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Application):
For Bodily Fluids
For Chemical Spills
Capacity (English):
0.33 gal.
0.49 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Capacity (English)
19-129-879 For Bodily Fluids 0.49 gal.
19-129-878 For Chemical Spills 0.33 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-129-879 Supplier Brady™ SPC™ Absorbents Supplier No. SKHAZQRKBF5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The HAZWIK™ Quick Response Kit provides fast and effective cleanup for emergency spills in a convenient, grab-and-go-style bag.

  • It contains everything you need for emergency spills in a convenient, grab-and-go-style bag.
  • Compact, single-use kit ideal for use in schools, hospitals and on janitorial carts.
  • Quickly cleans up hazardous bodily fluid spills: blood, vomit and urine.
  • Visual instructions on the package.
  • Contains PPE, tools, absorbent pads, organic granular, wipes and disposable materials.
  • Five individual kits per case.

Specifications

Color Green
Description Ideal for use in schools, hospitals and on janitorial carts
Capacity (English) 0.49 gal.
Material Polyethylene
Type Emergency Spill Kit
For Use With (Application) For Bodily Fluids
Includes 7.5 x 10 in. Pads, 6; Pair of Nitrile Gloves, 1; Bag Organic Coconut Coir Granular, 1; Scoop and Scraper, 1; Disposal Bag, 1; Cable Tie, 1; Surface Sanitizing Wipe, 1; Hand Sanitizing Wipe, 1
Width (English) 9 in.
Height (English) 13.13 in.
Depth (English) 19 in.
Weight (English) 0.4 lb.
Weight (Metric) 0.88 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.