Learn More
Cole-Parmer Jenway 6850 Double-Beam Spectrophotometer w/ Variable Bandwidth, IQ/OQ; 230 VAC
Supplier:  Cole-Parmer 8307008
Encompass_Preferred
This double beam, scanning, UV/Visible spectrophotometer has a variety of programmed measurement modes for photometrics, concentration, multi-wavelength, spectrum scanning, kinetics, quantitation, DNA and protein analysis making it ideal for a number of applications. The stable optics and two detectors measure your samples and reference simultaneously providing superior measurement accuracy. The variable spectral bandwidth provides your experiments with robust detection and quantitation limits, as well as optimal data precision and repeatability. This flexible platform allows the user to comply with multiple regulatory agencies using one spectrophotometer- saving money and time. Three scan speeds are available with wavelength scan intervals of 0.1, 0.2, 1, 2 or 5 nm. The large sample chamber is outfitted with a 10 x 10 mm cuvette holder, but allows for an 8-position cuvette changer (sold separately).
Catalog No. 50-196-1226
Add to Cart
Request a Quote