Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ SPC™ Hazwik™ Attack Pac™ Portable Spill Kit

Catalog No. 19819463 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
7 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-819-463 7 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-463 Supplier Brady™ Supplier No. SKHATK
Add to Cart
Add to Cart

Includes

  • 38.1 x 48.3cm (15 x 19 in.) Pads - 15
  • 7.62cm x 1.22m (3 in. x 4 ft.) SOCs - 3
  • Pair Nitrile Gloves - 1
  • Disposal Bag - 1
  • Goggles
  • Instruction Sheet

Efficient and space-saving single use kit for use with spills of hazardous materials.

  • Packed in a puncture- and UV-resistant foil bag
  • Includes grommet for hanging
  • Shipping wt: 11.34kg (25 lbs.)

Specifications

Capacity (English) 7 gal.
Type Portable Spill Kit
Includes (15) 15 x 19 in. pads, (3) 3 in. x 4 ft. socks, disposal bag, goggles, handbook, Nitrile gloves
Dimensions (L x W x H) 4 x 16 x 20 in.
Weight (English) 21.5 lb.
Weight (Metric) 47.41 kg
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.