Learn More
Brady™ SPC™ MAXX Enhanced Heavy Weight Oil Rolls
Catalog No. 17111189 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
70 gal./Bale
72 gal./Bale
Dimensions (L x W):
144 ft. x 19 in.
144 ft. x 38 in
Packaging Quantity:
1 Ea.
2/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W) Packaging Quantity
17-111-189 72 gal./Bale 144 ft. x 19 in. 2/Cs.
17-111-175 70 gal./Bale 144 ft. x 38 in 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-111-189 Supplier Brady™ Supplier No. SPC152
  • $333.00
    $232.65 / Case of 2
    Save $100.35
    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
For use with oil and petroleum based fluids
  • Single-ply construction with MAXX technology
  • Hydrophobic nature for outdoor applications
  • Floats indefinitely even when saturated with oil
  • Dimpled and perforated
  • Heavier and bonded to reduce linting
  • Available in various sizes
  • Specifications
    Capacity (English) 72 gal./Bale
    Material Polypropylene
    Type Oil Absorbent Roll
    For Use With (Application) Overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge Ponds
    Packaging Quantity 2/Cs.
    Dimensions (L x W) 144 ft. x 19 in.