Learn More
Brady™ SPC Premium Sorbent Boom
Catalog No. 19103110 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
61 gal.
Catalog No. Capacity (English)
19-103-110 61 gal.
1 options
Catalog No. 19-103-110 Supplier Brady™ Supplier No. SPC5510
  • $284.00
    $203.65 / Case of 2
    Save $80.35
    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Control spills in outfalls and discharge ponds. The Brady™ SPC Sorbent Booms are strong, lint-free, and gapless. They are easy to handle even when saturated with oil and petroleum based fluids.
  • Strong, lint-free and gapless
  • They are easy to handle even when saturated with oil and petroleum based fluids
  • Double 5” boom lies flat to cover more surface area
Specifications
Absorbancy 12.1 gl/gs
Capacity (English) 61 gal.
Material Polypropylene
Type Marine Sorbent Boom, Oil-only
Packaging Quantity 2/Cs.
Dimensions (L x W) 5 in. x 10 ft.