Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ SPC Universal Plus Rolls

Catalog No. 19003490 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
39 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-003-490 39 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-003-490 Supplier Brady™ Supplier No. SPCUN1019
  • $429.00
    $312.65 / Case of 2

    Save $116.35

    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Universal Plus products are chemically inert and green in color (the universal symbol for safety) to indicate that they're intended for use with hazardous materials.

Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert. Medium weight - optimal for most general maintenance needs. Universal Plus products feature 3-ply construction increasing durability while reducing lint. Perforations provide cost control and reduce waste by allowing you to use only what you need.

Specifications

Color Green
Capacity (English) 39 gal.
Capacity (Metric) 147.6 L
Material Polypropylene
Type Universal Plus Roll
For Use With (Application) Overspray leaks and drips, workstations, heavy clean-up and wipe-up , drums - liquid storage, outfalls and discharge ponds
Thickness 3-ply
Dimensions (L x W) 150 ft. x 15 in. (45.72 m x 38 cm)
Length (English) 150 ft.
Length (Metric) 45.72 m
Width (English) 15 in.
Width (Metric) 38 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.