Learn More
Aviva Systems Biology Corp SPC25 Antibody - middle region (ARP61828_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61828_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPC25. It was validated on Western Blot. . Alternate names: AD024, MGC22228, SPBC25, hSpc25ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-481