Learn More
Abcam SpCas9 D10A Nickase (Active), 1MG
Catalog No. 502703288
Encompass_Preferred
SpCas9 D10A Nickase (Active) is a Streptococcus pyogenes serotype M1 Full Length protein, in the 2 to 1368 aa range, expressed in Escherichia coli, with >=95% purity, < 10 EU/mg endotoxin level and suitable for SDS-PAGE, HPLC, Biological Activity.
Protein/ Peptide/ Enzyme Synonyms: csn1, SPy_1046, cas9, CRISPR-associated endonuclease Cas9/Csn1, SpCas9, SpyCas9
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-3288 Supplier Abcam Supplier No. AB3161721MG
SpCas9 D10A Nickase (Active) is a Streptococcus pyogenes serotype M1 Full Length protein, in the 2 to 1368 aa range, expressed in Escherichia coli, with >=95% purity, < 10 EU/mg endotoxin level and suitable for SDS-PAGE, HPLC, Biological Activity.
Protein/ Peptide/ Enzyme Synonyms: csn1, SPy_1046, cas9, CRISPR-associated endonuclease Cas9/Csn1, SpCas9, SpyCas9
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement