Learn More
Aldevron SpCas9 Nuclease, 0.25mg
Catalog No. 503073868
Encompass_Preferred
0.25mg dispense of SpCas9 Nuclease. Aldevron provides Research-grade Cas9 nucleases via Fisher Scientific for use in development work as well as standard cGMP products for clinical studies when ordered direct.
Functional Performance
•Ready to use for electroporation or transfection
•Extensive QC panels
•10 mg/mL formulation
Catalog No. 50-307-3868 Supplier Aldevron Supplier No. 92120.25MG
0.25mg dispense of SpCas9 Nuclease. Aldevron provides Research-grade Cas9 nucleases via Fisher Scientific for use in development work as well as standard cGMP products for clinical studies when ordered direct.
Functional Performance
•Ready to use for electroporation or transfection
•Extensive QC panels
•10 mg/mL formulation