Learn More
Aldevron SpCas9 Nuclease, 5mg
Catalog No. 503073869
Encompass_Preferred
5mg dispense of SpCas9 Nuclease. Aldevron provides Research-grade Cas9 nucleases via Fisher Scientific for use in development work as well as standard cGMP products for clinical studies when ordered direct.
Functional Performance
•Ready to use for electroporation or transfection
•Extensive QC panels
•10 mg/mL formulation
Catalog No. 50-307-3869 Supplier Aldevron Supplier No. 92125MG
Add to Cart
Add to Cart
Request a Quote
5mg dispense of SpCas9 Nuclease. Aldevron provides Research-grade Cas9 nucleases via Fisher Scientific for use in development work as well as standard cGMP products for clinical studies when ordered direct.
Functional Performance
•Ready to use for electroporation or transfection
•Extensive QC panels
•10 mg/mL formulation