Learn More
Chemglass Life Sciences 24/40 Inner Joint, 16in OAL, on 28mm OD x 2mm W tubing
SDP
Supplier:  Chemglass Life Sciences CG10904
Encompass_Preferred
24/40 Inner Joint, 16in OAL, on 28mm OD x 2mm W tubing. Standard taper inner and outer lengths tooled on longer lengths or different sizes than normally available under part numbers CG-100 through CG-105. These joints offer the glassblower the opportunity to construct apparatus made of heavier tubing or without unnecessary seals.
Catalog No. CG10904