Learn More
Chemglass Life Sciences 24/40 Inner Joint, 16in OAL, on 22mm OD x 1.8mm W tubing
SDP
Catalog No. CG10903
Encompass_Preferred
24/40 Inner Joint, 16in OAL, on 22mm OD x 1.8mm W tubing. Standard taper inner and outer lengths tooled on longer lengths or different sizes than normally available under part numbers CG-100 through CG-105. These joints offer the glassblower the opportunity to construct apparatus made of heavier tubing or without unnecessary seals.
Catalog No. CG10903 Supplier Chemglass Life Sciences Supplier No. CG10903
24/40 Inner Joint, 16in OAL, on 22mm OD x 1.8mm W tubing. Standard taper inner and outer lengths tooled on longer lengths or different sizes than normally available under part numbers CG-100 through CG-105. These joints offer the glassblower the opportunity to construct apparatus made of heavier tubing or without unnecessary seals.