Learn More
Chemglass Life Sciences 24/40 Outer Joint, 16in OAL, on 25mm OD x 1.5mm W tubing
SDP
Catalog No. CG10928
Encompass_Preferred
24/40 Outer Joint, 16in OAL, on 25mm OD x 1.5mm W tubing. Standard taper inner and outer lengths tooled on longer lengths or different sizes than normally available under part numbers CG-100 through CG-105. These joints offer the glassblower the opportunity to construct apparatus made of heavier tubing or without unnecessary seals.
Catalog No. CG10928 Supplier Chemglass Life Sciences Supplier No. CG10928
24/40 Outer Joint, 16in OAL, on 25mm OD x 1.5mm W tubing. Standard taper inner and outer lengths tooled on longer lengths or different sizes than normally available under part numbers CG-100 through CG-105. These joints offer the glassblower the opportunity to construct apparatus made of heavier tubing or without unnecessary seals.