Learn More
Chemglass Life Sciences 34/45 Outer Joint, 16in OAL, on 38mm OD x 2mm W tubing
SDP
Catalog No. CG10946
Encompass_Preferred
34/45 Outer Joint, 16in OAL, on 38mm OD x 2mm W tubing. Standard taper inner and outer lengths tooled on longer lengths or different sizes than normally available under part numbers CG-100 through CG-105. These joints offer the glassblower the opportunity to construct apparatus made of heavier tubing or without unnecessary seals.
Catalog No. CG10946 Supplier Chemglass Life Sciences Supplier No. CG10946
34/45 Outer Joint, 16in OAL, on 38mm OD x 2mm W tubing. Standard taper inner and outer lengths tooled on longer lengths or different sizes than normally available under part numbers CG-100 through CG-105. These joints offer the glassblower the opportunity to construct apparatus made of heavier tubing or without unnecessary seals.