Learn More
Chemglass Life Sciences 45/50 Drip Joint, 16in OAL, on 41mm OD x 2mm W tubing
SDP
Catalog No. CG10911
Encompass_Preferred
45/50 Drip Joint, 16in OAL, on 41mm OD x 2mm W tubing. Standard taper inner and outer lengths tooled on longer lengths or different sizes than normally available under part numbers CG-100 through CG-105. These joints offer the glassblower the opportunity to construct apparatus made of heavier tubing or without unnecessary seals.
Catalog No. CG10911 Supplier Chemglass Life Sciences Supplier No. CG10911
45/50 Drip Joint, 16in OAL, on 41mm OD x 2mm W tubing. Standard taper inner and outer lengths tooled on longer lengths or different sizes than normally available under part numbers CG-100 through CG-105. These joints offer the glassblower the opportunity to construct apparatus made of heavier tubing or without unnecessary seals.