Learn More
Chemglass Life Sciences 55/50 Outer Joint, 16in OAL, on 54mm OD x 2.4mm W tubing
SDP
Catalog No. CG10950
Encompass_Preferred
55/50 Outer Joint, 16in OAL, on 54mm OD x 2.4mm W tubing. Standard taper inner and outer lengths tooled on longer lengths or different sizes than normally available under part numbers CG-100 through CG-105. These joints offer the glassblower the opportunity to construct apparatus made of heavier tubing or without unnecessary seals.
Catalog No. CG10950 Supplier Chemglass Life Sciences Supplier No. CG10950
55/50 Outer Joint, 16in OAL, on 54mm OD x 2.4mm W tubing. Standard taper inner and outer lengths tooled on longer lengths or different sizes than normally available under part numbers CG-100 through CG-105. These joints offer the glassblower the opportunity to construct apparatus made of heavier tubing or without unnecessary seals.