Learn More
Newcomer Supply Specimen Chuck w/Colored O-ring & Orientation Mark, Round, 30 mm, Red
SDP
Catalog No. NC1841642
Encompass
The 30 mm specimen chuck has a colored o-ring and oriention mark. It has proper tolerance and soft aluminum. It is suitable for Leica (except new CM1950), TBS, Tanner, Avantik and other common cryostats. Available in Blue, Green, Orange, Pink, Dark Purple, Red, White and Yellow.
Catalog No. NC1841642 Supplier Newcomer Supply Supplier No. 6865R
The 30 mm specimen chuck has a colored o-ring and oriention mark. It has proper tolerance and soft aluminum. It is suitable for Leica (except new CM1950), TBS, Tanner, Avantik and other common cryostats. Available in Blue, Green, Orange, Pink, Dark Purple, Red, White and Yellow.