Learn More
Newcomer Supply Specimen Chuck w/Colored O-ring & Orientation Mark, Round, 30 mm, Yellow
SDP
Catalog No. NC1841644
Encompass
The 30 mm specimen chuck has a colored o-ring and oriention mark. It has proper tolerance and soft aluminum. It is suitable for Leica (except new CM1950), TBS, Tanner, Avantik and other common cryostats. Available in Blue, Green, Orange, Pink, Dark Purple, Red, White and Yellow.
Catalog No. NC1841644 Supplier Newcomer Supply Supplier No. 6865Y
The 30 mm specimen chuck has a colored o-ring and oriention mark. It has proper tolerance and soft aluminum. It is suitable for Leica (except new CM1950), TBS, Tanner, Avantik and other common cryostats. Available in Blue, Green, Orange, Pink, Dark Purple, Red, White and Yellow.