Learn More
Electron Microscopy Sciences Specimen Containers w/Snap Cap, 1000 ml, 140 mm height, 111 mm Opening
SDP
Catalog No. 501931653
Encompass_Preferred
These disposable containers are ideal for collection and storage of fluids, powders, solids, pathology specimens and hazardous samples. Lids are tight fitting. They resist temperatures form -20 to + 80 degrees Celcius. The container, cap and closure are made of polypropylene.
Capacity: 1000 ml
Height: 140 mm
Opening: 111 mm
Catalog No. 50-193-1653 Supplier Electron Microscopy Sciences Supplier No. 6423903
These disposable containers are ideal for collection and storage of fluids, powders, solids, pathology specimens and hazardous samples. Lids are tight fitting. They resist temperatures form -20 to + 80 degrees Celcius. The container, cap and closure are made of polypropylene.
Capacity: 1000 ml
Height: 140 mm
Opening: 111 mm