Learn More
Electron Microscopy Sciences Specimen Containers w/Snap Cap, 300 ml, 45 mm height, 111 mm Opening
SDP
Catalog No. 501931651
Encompass_Preferred
These disposable containers are ideal for collection and storage of fluids, powders, solids, pathology specimens and hazardous samples. Lids are tight fitting. They resist temperatures form -20 to + 80 degrees Celcius. The container, cap and closure are made of polypropylene.
Capacity: 300 ml
Height: 45 mm
Opening: 111 mm
Catalog No. 50-193-1651 Supplier Electron Microscopy Sciences Supplier No. 6423901
These disposable containers are ideal for collection and storage of fluids, powders, solids, pathology specimens and hazardous samples. Lids are tight fitting. They resist temperatures form -20 to + 80 degrees Celcius. The container, cap and closure are made of polypropylene.
Capacity: 300 ml
Height: 45 mm
Opening: 111 mm