Learn More
Azzota Corp Visible Spectrophotometer, 6Nm Bandpass, 360-1000 Nm. Accuracy 1.0%T, Manual Wl Setting
Catalog No. 501179616
Encompass_Preferred
Visible Spectrophotometer, 6nm Bandpass, 360-1000 nm. Accuracy 1.0%T, Manual WL setting
Catalog No. 50-117-9616 Supplier Azzota Corp Supplier No. SV1000
Visible Spectrophotometer, 6nm Bandpass, 360-1000 nm. Accuracy 1.0%T, Manual WL setting