Learn More
Medchemexpress LLC SPD-473 citrate | 161190-26-7 | 99.8% | 552.47 g/mol | C23H31Cl2NO8S | 5 MG
SDP
Catalog No. 5000263219
Encompass_Preferred
SPD-473 citrate is the citrate salt of a small-molecule triple reuptake inhibitor that blocks serotonin, dopamine, and norepinephrine transport. It is supplied as a solid research chemical for in vitro and in vivo neuroscience and pharmacology studies and is accompanied by analytical and safety documentation.
  • Triple reuptake inhibitor activity for serotonin, dopamine, and norepinephrine.
  • Citrate salt form supplied as a solid.
  • Supplied in small research quantities (1-100 mg) suitable for laboratory studies.
  • High purity: 99.8%.
  • Analytical data and documentation available, including data sheet, SDS, COA, HNMR, and LCMS.
Catalog No. 50-002-63219 Supplier Medchemexpress LLC Supplier No. HY1016125MG
SPD-473 citrate is the citrate salt of a small-molecule triple reuptake inhibitor that blocks serotonin, dopamine, and norepinephrine transport. It is supplied as a solid research chemical for in vitro and in vivo neuroscience and pharmacology studies and is accompanied by analytical and safety documentation.
  • Triple reuptake inhibitor activity for serotonin, dopamine, and norepinephrine.
  • Citrate salt form supplied as a solid.
  • Supplied in small research quantities (1-100 mg) suitable for laboratory studies.
  • High purity: 99.8%.
  • Analytical data and documentation available, including data sheet, SDS, COA, HNMR, and LCMS.