Learn More
Medchemexpress LLC SPD-473 citrate | 161190-26-7 | MFCD31382144 | 99.8% | 552.47 g/mol | C23H31Cl2NO8S | 1 MG
SDP
Catalog No. 5000248065
Encompass_Preferred
SPD-473 citrate is the citrate salt of a triple reuptake inhibitor that blocks serotonin, dopamine, and norepinephrine transporters. Supplied for research use, it is a characterized white to off-white solid with documented purity and solubility data for in vitro and in vivo studies.
  • Acts as a serotonin, dopamine, and norepinephrine reuptake inhibitor.
  • High purity (99.8%).
  • Molecular weight 552.47 g/mol.
  • White to off-white solid suitable for laboratory handling.
  • High solubility in DMSO (100 mg/mL); in vivo formulation protocols provided.
  • Available in 1 mg, 5 mg, 10 mg, and 25 mg pack sizes for screening and pilot studies.
Catalog No. 50-002-48065 Supplier Medchemexpress LLC Supplier No. HY1016121MG
SPD-473 citrate is the citrate salt of a triple reuptake inhibitor that blocks serotonin, dopamine, and norepinephrine transporters. Supplied for research use, it is a characterized white to off-white solid with documented purity and solubility data for in vitro and in vivo studies.
  • Acts as a serotonin, dopamine, and norepinephrine reuptake inhibitor.
  • High purity (99.8%).
  • Molecular weight 552.47 g/mol.
  • White to off-white solid suitable for laboratory handling.
  • High solubility in DMSO (100 mg/mL); in vivo formulation protocols provided.
  • Available in 1 mg, 5 mg, 10 mg, and 25 mg pack sizes for screening and pilot studies.