Learn More
TARGETMOL CHEMICALS INC SPD304 DIHYDROCHLORIDE 5MG
SDP
Catalog No. 502581391
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SPD304 dihydrochloride is a selective inhibitor of TNF-(alpha) with an IC50 of 22 µM. SPD304 dihydrochloride promotes dissociation of TNF trimers. purity: 98%
Catalog No. 50-258-1391 Supplier TARGETMOL CHEMICALS INC Supplier No. T129855MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SPD304 dihydrochloride is a selective inhibitor of TNF-(alpha) with an IC50 of 22 µM. SPD304 dihydrochloride promotes dissociation of TNF trimers. purity: 98%