Learn More
TARGETMOL CHEMICALS INC SPDB 10MG
SDP
Catalog No. 502576775
Encompass_Preferred
Also available in 2 mg 5 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SPDB is a small fragment linked to DM4 conjugates. It conjugates to antibodies utilizing disulfide linkers anti-EGFR-SPDB-DM4 has been widely used. purity: 95%
Catalog No. 50-257-6775 Supplier TARGETMOL CHEMICALS INC Supplier No. T434610MG
Also available in 2 mg 5 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SPDB is a small fragment linked to DM4 conjugates. It conjugates to antibodies utilizing disulfide linkers anti-EGFR-SPDB-DM4 has been widely used. purity: 95%