Learn More
Aviva Systems Biology Corp SPDEF Antibody - N-terminal region (ARP31841_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31841_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SPDEF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SPDEF. Alternate names: PDEF, RP11-375E1__A.3, bA375E1.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54338