Learn More
Aviva Systems Biology Corp SPDEF Antibody - middle region (ARP32533_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32533_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPDEF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SPDEF. Alternate names: PDEF, RP11-375E1__A.3, bA375E1.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54638