Learn More
Aviva Systems Biology Corp SPDEF Antibody - C-terminal region (ARP38992_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38992_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPDEF. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human SPDEF. Alternate names: PDEF, RP11-375E1__A.3, bA375E1.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-771