Learn More
Medchemexpress LLC Spdmv | 890409-85-5 | 96.4% | 354.44 g/mol | C15H18N2O4S2 | 250 MG
SDP
Catalog No. 5000202607
Encompass_Preferred
SPDMV is a glutathione-cleavable antibody-drug conjugate (ADC) linker provided for research use. It releases payloads under reducing (glutathione) conditions and is used to prepare cleavable ADCs in laboratory research. CAS 890409-85-5; molecular weight 354.44 g/mol; formula C15H18N2O4S2; typical purity ~96.4%.
  • Glutathione cleavable linker for controlled payload release.
  • Suitable for construction of cleavable ADCs.
  • Provided for research use only (not for human use).
  • Available in milligram quantities, including 50 MG, 100 MG, and 250 MG.
  • High purity typical of supplied lots (~96.4%).
Catalog No. 50-002-02607 Supplier Medchemexpress LLC Supplier No. HY129368250MG
SPDMV is a glutathione-cleavable antibody-drug conjugate (ADC) linker provided for research use. It releases payloads under reducing (glutathione) conditions and is used to prepare cleavable ADCs in laboratory research. CAS 890409-85-5; molecular weight 354.44 g/mol; formula C15H18N2O4S2; typical purity ~96.4%.
  • Glutathione cleavable linker for controlled payload release.
  • Suitable for construction of cleavable ADCs.
  • Provided for research use only (not for human use).
  • Available in milligram quantities, including 50 MG, 100 MG, and 250 MG.
  • High purity typical of supplied lots (~96.4%).