Learn More
Medchemexpress LLC HY-100216 500mg Medchemexpress, SPDP CAS:68181-17-9 Purity:>98%
SDP
Catalog No. 501931756
Encompass_Preferred
Medchemexpress, HY-100216 500mg SPDP CAS:68181-17-9 SPDP is a short-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1756 Supplier Medchemexpress LLC Supplier No. HY100216500MG
Medchemexpress, HY-100216 500mg SPDP CAS:68181-17-9 SPDP is a short-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.