Learn More
TARGETMOL CHEMICALS INC SPDP 50MG
SDP
Catalog No. 502576466
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. SPDP is a short-chain crosslinker which is used for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls. SPDP could activate equal amounts of anti-CD11c and anti-CTLA-4 Abs (in borate buffered saline; pH 8.5). purity: 100%
Catalog No. 50-257-6466 Supplier TARGETMOL CHEMICALS INC Supplier No. T674150MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. SPDP is a short-chain crosslinker which is used for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls. SPDP could activate equal amounts of anti-CD11c and anti-CTLA-4 Abs (in borate buffered saline; pH 8.5). purity: 100%