Learn More
Medchemexpress LLC SPDP-C6-NHS ester | 158913-22-5 | 98.1% | 425.52 g/mol | C18H23N3O5S2 | 250MG
SDP
Catalog No. 5000239664
Encompass_Preferred
SPDP-C6-NHS ester is an NHS-activated, cleavable linker that contains a disulfide bond and is used for bioconjugation and PROTAC synthesis. The reagent couples to primary amines via its NHS ester and enables reversible linkage through disulfide reduction. It is supplied as a powder with specified storage and stability conditions and is intended for research use only.
  • Cleavable disulfide linker for reversible conjugation
  • NHS ester reactive toward primary amines
  • Suitable for PROTAC synthesis and bioconjugation applications
  • High purity (98.1% as listed)
  • Available in multiple pack sizes, including 250 mg
  • Powder storage recommendations: -20°C for long term, 4°C for shorter term
Catalog No. 50-002-39664 Supplier Medchemexpress LLC Supplier No. HY124377250MG
SPDP-C6-NHS ester is an NHS-activated, cleavable linker that contains a disulfide bond and is used for bioconjugation and PROTAC synthesis. The reagent couples to primary amines via its NHS ester and enables reversible linkage through disulfide reduction. It is supplied as a powder with specified storage and stability conditions and is intended for research use only.
  • Cleavable disulfide linker for reversible conjugation
  • NHS ester reactive toward primary amines
  • Suitable for PROTAC synthesis and bioconjugation applications
  • High purity (98.1% as listed)
  • Available in multiple pack sizes, including 250 mg
  • Powder storage recommendations: -20°C for long term, 4°C for shorter term