Learn More
Abcam SPDP crosslinker, Cysteine and primary amine reactive short chain crosslinker, 50MG
Catalog No. 502705109
Encompass_Preferred
MW 312.4 Da, Purity >90%. Cysteine and primary amine reactive short chain crosslinker. Induces amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5109 Supplier Abcam Supplier No. AB14561350MG
MW 312.4 Da, Purity >90%. Cysteine and primary amine reactive short chain crosslinker. Induces amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls.
The product is subject to the following: Abcam Restricted Use Statement