Learn More
Medchemexpress LLC SPDP-PEG24-NHS ester | 95.0% | C63H113N3O29S2 | 25 MG
SDP
Catalog No. 5000352529
Encompass_Preferred
SPDP-PEG24-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs, which are molecules designed to selectively degrade target proteins by leveraging the intracellular ubiquitin-proteasome system. PROTACs consist of two ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other targeting a protein of interest. This product is for research use only and not intended for medical applications or sold to patients.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs leverage the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-52529 Supplier Medchemexpress LLC Supplier No. 25MGHY141357
SPDP-PEG24-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs, which are molecules designed to selectively degrade target proteins by leveraging the intracellular ubiquitin-proteasome system. PROTACs consist of two ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other targeting a protein of interest. This product is for research use only and not intended for medical applications or sold to patients.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs leverage the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only