Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SPDP-PEG4-acid | 581065-97-6 | C19H30N2O7S2 | 50MG
SDP

Supplier:  Medchemexpress LLC HY14135250MG

Encompass_Preferred

SPDP-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting two ligands: one for an E3 ubiquitin ligase and another for the target protein.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Designed for selective protein degradation
  • Colorless to light yellow liquid appearance
  • Store at 4°C under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen

Catalog No. 50-003-53861


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.