Learn More
Medchemexpress LLC SPDP-PEG4-NHS ester | 1334177-95-5 | 95.0% | 100 MG
SDP
Catalog No. 5000339618
Encompass_Preferred
SPDP-PEG4-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules that contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. These compounds exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-39618 Supplier Medchemexpress LLC Supplier No. HY130195100MG
SPDP-PEG4-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules that contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. These compounds exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only