Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SPDP-PEG4-NHS ester | 1334177-95-5 | 95.0% | 50 MG
SDP

Catalog No. 5000339940
Encompass_Preferred

SPDP-PEG4-NHS ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules operate by utilizing the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins. PROTACs consist of two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, while the other targets the specific protein of interest.

  • For research use only.
  • Store pure form at -20°C for up to 3 years.
  • Store in solvent at -80°C for up to 6 months, or at -20°C for up to 1 month.
  • If stored at -80°C, stock solutions should be used within 6 months.
  • If stored at -20°C, stock solutions should be used within 1 month.

Catalog No. 50-003-39940 Supplier Medchemexpress LLC Supplier No. HY13019550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SPDP-PEG4-NHS ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules operate by utilizing the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins. PROTACs consist of two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, while the other targets the specific protein of interest.

  • For research use only.
  • Store pure form at -20°C for up to 3 years.
  • Store in solvent at -80°C for up to 6 months, or at -20°C for up to 1 month.
  • If stored at -80°C, stock solutions should be used within 6 months.
  • If stored at -20°C, stock solutions should be used within 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.