Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 6mL, 1/16 inch, 10m- 100/Pk

Catalog No. 502400054 Shop All Phenomenex Products
Encompass_Preferred

SPE 10 m Polyethylene Frits AS006-A

Catalog No. 50-240-0054 Supplier Phenomenex Inc Supplier No. AS006A

missing translation for 'validMainframeMsg'

SPE 10 m Polyethylene Frits AS006-A

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.