Learn More
MilliporeSigma™ Supelco™ SPE 96-Deep Square Well Collection Plate
Catalog No. 111003690 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric) Well:
2 mL
Catalog No. Volume (Metric) Well
11-100-3690 2 mL
1 options
Catalog No. 11-100-3690 Supplier MilliporeSigma™ Supelco™ Supplier No. 575653U
Add to Cart
Add to Cart
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
TRUSTED_SUSTAINABILITY
Specifications
Column Type SPE
For Use With (Application) Food and Beverages
Type 96-well Plate
Volume (Metric) Well 2 mL
Material Polypropylene
Quantity 50/Pk.